Polyethylene glycol 2,000
Polyethylene glycol 2,000
Polyethylene glycol 2,000
Polyethylene glycol 2,000
Thermo Scientific Chemicals
Polyethylene glycol 2,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Catalog NumberQuantity
FSKB22181.0B1000 g
Catalog number FSKB22181.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene glycol 2,000
pH5 to 7
CAS25322-68-3
Melting Point53°C to 55°C
Recommended StorageAmbient temperatures
View more
Polyethylene glycol is used in the field of pharmaceuticals, cosmetics, paper, rubber and ceramics and in wood processing. It is also used in the manufacture of surfactants, dispersant resin and plastics. It acts as a dye carrier in paints and inks; soldering fluxes with good spreading property and as a softener and antistatic agent for textiles. It is also one of the main materials of ester type surface active agent.