Polyethylene glycol 200
Polyethylene glycol 200
Polyethylene glycol 200
Polyethylene glycol 200
Thermo Scientific Chemicals
Polyethylene glycol 200
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Catalog NumberQuantity
FSKB21918.30250 g
Catalog number FSKB21918.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene glycol 200
CAS25322-68-3
Physical FormLiquid
Health Hazard 1H227-H303-H315-H320-H333-H335
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c
View more
Polyethylene glycol is used in electron microscopy. It is used as a solvent, a dye carrier in inks and a dispersant in water based coatings. Its derivatives like PEG esters are used as surfactants. In the textile industry, it is used as a lubricant, softener, dye carrier dispersant and an anti-static agent. It finds application in personal care products like skin creams, lotions and bath gels.