Periodic acid, 34% w/w aq. soln.
Periodic acid, 34% w/w aq. soln.
Periodic acid, 34% w/w aq. soln.
Periodic acid, 34% w/w aq. soln.
Thermo Scientific Chemicals
Periodic acid, 34% w/w aq. soln.
CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
Have Questions?
Catalog NumberQuantity
FSKB20172.36500 g
Catalog number FSKB20172.36
Price (SGD)
595.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
595.00
EA
Add to cart
Specifications
Chemical Name or MaterialPeriodic acid
CAS10450-60-9
Concentration34% w/w aq. soln.
Health Hazard 1H272-H290-H314-H335
Health Hazard 2GHS H Statement
H272-H314
May intensify fire; oxidizer.