Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Solvents
▸
Methanol
▸
Methanol (HPLC), Fisher Chemical™
Element Search
Structure Search
Thermo Scientific Chemicals
Methanol (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Have Questions?
Catalog Number
Quantity
Packaging
FSKA452-4
4 L
Glass Bottle
Catalog number
FSKA452-4
Price (SGD)
-
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Methanol
Cleanup Symbol
Solvent
Melting Point
-98°C
Absorbance
0.025 AU max. at 254nm, 0.15 AU max. at 230nm, 0.30 AU max. at 220nm, 1.00 AU max. at 205nm
CAS
67-56-1
View more