2-Amino-6-bromobenzothiazole, 98%
2-Amino-6-bromobenzothiazole, 98%
2-Amino-6-bromobenzothiazole, 98%
2-Amino-6-bromobenzothiazole, 98%
Thermo Scientific Chemicals
2-Amino-6-bromobenzothiazole, 98%
CAS: 15864-32-1 | C7H5BrN2S | 229.10 g/mol
Have Questions?
Catalog NumberQuantity
FSKA18554.065 g
Catalog number FSKA18554.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-6-bromobenzothiazole
CAS15864-32-1
Health Hazard 1H302-H315-H317-H319-H335
Health Hazard 2GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c