Nigrosin water soluble
Nigrosin water soluble
Nigrosin water soluble
Nigrosin water soluble
Thermo Scientific Chemicals
Nigrosin water soluble
CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol
Have Questions?
Catalog NumberQuantity
FSKA18147.22100 g
Catalog number FSKA18147.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNigrosin
Name Notewater soluble
CAS8005-03-6
Molecular FormulaC22H14N6Na2O9S2
MDL NumberMFCD00044681
View more
It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner?s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner′s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.
Solubility
Soluble in water, ethanol, oil, nigrosines and fat.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Zhang S.; Zhao XW.; Xu H.; Zhu R.; Gu ZZ. Fabrication of photonic crystals with nigrosine-doped poly(MMA-co-DVB-co-MAA) particles.J Colloid Interface Sci.2007,316(1), 168-74.
  2. J.P. Kaltenbach.; Merle H. Kaltenbach.; W.B. Lyons. Nigrosin as a dye for differentiating live and dead ascites cells.Experimental Cell Research,1958,15(1), 112-117.