Sodium sulfite, anhydrous, 98%
Sodium sulfite, anhydrous, 98%
Sodium sulfite, anhydrous, 98%
Sodium sulfite, anhydrous, 98%
Sodium sulfite, anhydrous, 98%
Thermo Scientific Chemicals
Sodium sulfite, anhydrous, 98%
CAS: 7757-83-7 | Na2O3S | 126.037 g/mol
Have Questions?
Catalog NumberQuantity
FSKA17933.36500 g
Catalog number FSKA17933.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium sulfite
Melting Point>500°C (decomposition)
Name Noteanhydrous
CAS7757-83-7
Recommended StorageAmbient temperatures
View more
Sodium sulfite is used in the photography industry to protect developing solutions from oxidation, in the pulp and paper industry, in water treatment as an oxygen scavenger agent, as a desulfurizing and dechlorinating agent in the leather industry and as a bleaching agent in textile industry. It is used as a preservative to prevent dried fruit from discoloring. It finds application as a sulfonation and sulfomethylation agent in the chemical industry. It is involved in the production of sodium thiosulfate. Other applications include forth flotation ores, oil recovery and making dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium sulfite is used in the photography industry to protect developing solutions from oxidation, in the pulp and paper industry, in water treatment as an oxygen scavenger agent, as a desulfurizing and dechlorinating agent in the leather industry and as a bleaching agent in textile industry. It is used as a preservative to prevent dried fruit from discoloring. It finds application as a sulfonation and sulfomethylation agent in the chemical industry. It is involved in the production of sodium thiosulfate. Other applications include forth flotation ores, oil recovery and making dyes.
Solubility
Soluble in water and glycerol. Insoluble in alcohol, liquid chlorine and liquid ammonia.
Notes
Hygroscopic. Moisture and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Cavka, A.; Martín, C.; Alriksson, B.; Mörtsell, M.; Jönsson, L. J. Techno-economic evaluation of conditioning with sodium sulfite for bioethanol production from softwood. Bioresour. Technol. 2015, 196, 129-135.
  2. Du, F.; Alam, M. N.; Pawliszyn, J. Aptamer-functionalized solid phase microextraction-liquid chromatography/tandem mass spectrometry for selective enrichment and determination of thrombin. Anal. Chim. Acta. 2014, 845, 45-52.