Sodium thiosulfate pentahydrate, 99+%
Sodium thiosulfate pentahydrate, 99+%
Sodium thiosulfate pentahydrate, 99+%
Sodium thiosulfate pentahydrate, 99+%
Thermo Scientific Chemicals
Sodium thiosulfate pentahydrate, 99+%
CAS: 10102-17-7 | H10Na2O8S2 | 248.172 g/mol
Have Questions?
Catalog NumberQuantity
FSKA17914.36500 g
Catalog number FSKA17914.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point100°C (in -5H2O)
Chemical Name or MaterialSodium thiosulfate pentahydrate
Melting Point48°C
CAS10102-17-7
Health Hazard 1H500
View more
Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivative S-alkylthiosulfonates are called Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solution and to check pH of bleach solutions with liquid indicators. Other used include tanning of leather, as an active component in hand warmers, in chemical heating pads and as an anionic surfactant to aid in dispersion of pharmaceutical preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivative S-alkylthiosulfonates are called Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solution and to check pH of bleach solutions with liquid indicators. Other used include tanning of leather, as an active component in hand warmers, in chemical heating pads and as an anionic surfactant to aid in dispersion of pharmaceutical preparations.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Incompatible with sodium nitrate, acids, strong oxidizing agents, iodides, silver salts, mercury salts and lead.
RUO – Research Use Only
General References:
  1. Moral-Vico, J.; Sánchez-Redondo, S.; Lichtenstein, M. P.; Suñol, C.; Casañ-Pastor, N. Nanocomposites of iridium oxide and conducting polymers as electroactive phases in biological media. Acta Biomater. 2014, 10 (5), 2177-2186.
  2. James, T. H.; Cannon, C.; Apblett, A.; Materer, N. F. Sodium Dithionite Purity and Decomposition Products in Solid Samples Spanning 50 Years. Phosphorus Sulfur Silicon Relat. Elem. 2015, 190 (2), 158-169.