Tartrazine
Tartrazine
Tartrazine
Thermo Scientific Chemicals
Tartrazine
C.I. 19140 | CAS: 1934-21-0 | C16H9N4Na3O9S2 | 534.356 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSKA17682.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTartrazine
CAS1934-21-0
Health Hazard 1H317-H334
Health Hazard 2GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Health Hazard 3P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
View more
It can be employed as a biological stain and a food colorant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be employed as a biological stain and a food colorant.
Solubility
Soluble in water 260 g/L (30°C).
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only
General References:
  1. Tran CM.; Fujita N.; Huang BL.; Ong JR.; Lyons KM.; Shapiro IM.; Risbud MV. Hypoxia-inducible factor (HIF)-1α and CCN2 form a regulatory circuit in hypoxic nucleus pulposus cells: CCN2 suppresses HIF-1α level and transcriptional activity.J Biol Chem.2013,288(18),12654-12666.
  2. Zhou F.; Xin H.; Liu T.; Li GY.; Gao ZZ, Liu J.; Li WR.; Cui WS.; Bai GY.; Park NC.; Xin ZC. Effects of icariside II on improving erectile function in rats with streptozotocin-induced diabetes.J Androl.2012,33(5), 832-844.