Polyethylene glycol 1,500
Polyethylene glycol 1,500
Polyethylene glycol 1,500
Polyethylene glycol 1,500
Thermo Scientific Chemicals
Polyethylene glycol 1,500
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Catalog NumberQuantity
FSKA16241.30250 g
Catalog number FSKA16241.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene glycol 1,500
pH5 to 7
CAS25322-68-3
Melting Point44°C to 48°C
Recommended StorageAmbient temperatures
View more
Cell fusion induced by polyethylene glycol(PEG) has become a standard method in somatic cell genetics. PEG promoted cell fusion is involved in the production of hybridoma cells.