Triton X-100
Triton X-100
Triton X-100
Thermo Scientific Chemicals
Triton X-100
CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSKA16046.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTriton X-100
CAS9002-93-1
Health Hazard 1H302-H318
Boiling Point>250°C
Health Hazard 3P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
View more
Detergent, equivalent to Nonidet P-40.Triton™ X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Detergent, equivalent to Nonidet P-40.Triton|r X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Nonionic detergent and emulsifier.
  2. El-Agamey, A.; El-Hagrasy, M. A.; Suenobu, T.; Fukuzumi, S. Influence of pH on the decay of beta-carotene radical cation in aqueous Triton X-100: A laser flash photolysis study. J. Photochem. Photobiol., B 2015, 146, 68-73.
  3. Patel, V.; Bharatiya, B.; Ray, D.; Aswal, V. K.; Bahadur, P. Investigations on microstructural changes in pH responsive mixed micelles of Triton X-100 and bile salt. J. Colloid Interface Sci. 2015, 441, 106-112.