Solvent Blue 38
Solvent Blue 38
Solvent Blue 38
Thermo Scientific Chemicals
Solvent Blue 38
CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol
Have Questions?
Catalog NumberQuantity
FSKA15395.1425 g
Catalog number FSKA15395.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFast Blue
Melting Point∼235°C (decomposition)
CAS1328-51-4
Recommended StorageAmbient temperatures
EINECS Number215-523-1
View more
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.
Solubility
Soluble in water (partly), and methanol (1 mg/ml).
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. L Novikova; L Novikov; J.-O Kellerth. Persistent neuronal labeling by retrograde fluorescent tracers: a comparison between Fast Blue, Fluoro-Gold and various dextran conjugates. Journal of Neuroscience Methods. 1997, 74 (1), 9-15.
  2. Margolis G, pickett JP. New applications of the Luxol fast blue myelin stain. Lab Invest. 1956, 5 (6), 459-74.