n-Decane, 99%
n-Decane, 99%
n-Decane, 99%
Thermo Scientific Chemicals
n-Decane, 99%
CAS: 124-18-5 | C10H22 | 142.286 g/mol
Have Questions?
Catalog NumberQuantity
FSKA14732.AE100 mL
Catalog number FSKA14732.AE
Price (SGD)
98.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
98.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Decane
CAS124-18-5
Health Hazard 1H226-H304
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
Melting Point°C to 30°C
View more
n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).
Solubility
Soluble in alcohol and ethers. Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ji, N.; Wang, X.; Weidenthaler, C.; Spliethoff, B.; Rinaldi, R. Iron(II) Disulfides as Precursors of Highly Selective Catalysts for Hydrodeoxygenation of Dibenzyl Ether into Toluene. Chem. Cat. Chem. 2015, 7 (6), 960-966.
  2. Khoshsima, A.; Dehghani, M. R.; Touraud, D.; Kunz, W. An investigation of the fish diagrams of water or brine/decane or dodecane/propylene glycol ether (C3P1 or C3P2) systems. J. Mol. Liq. 2015, 206, 170-175.