2-Amino-5-bromopyridine, 98%
2-Amino-5-bromopyridine, 98%
2-Amino-5-bromopyridine, 98%
2-Amino-5-bromopyridine, 98%
Thermo Scientific Chemicals
2-Amino-5-bromopyridine, 98%
CAS: 1072-97-5 | C5H5BrN2 | 173.013 g/mol
Have Questions?
Catalog NumberQuantity
FSKA14280.065 g
Catalog number FSKA14280.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-bromopyridine
CAS1072-97-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.
Solubility
Soluble in methanol, chloroform, ethyl acetate. Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Light sensitive. Store under argon.
RUO – Research Use Only
General References:
  1. Antonio Luque; Jon Sertucha; Luis Lezam; Teófilo Rojo; and Pascual Román. Synthesis, characterisation and crystal structure of 2-aminopyridinium(2-amino-5-bromopyridine)tribromocuprate(II)and bis(2-aminopyridinium)tetrabromocuprate(II). J. Chem. Soc., Dalton Trans. 1997, (5), 847-854.
  2. N. Sundaraganesan; H. Saleem; S. Mohan. Vibrational spectra, assignments and normal coordinate analysis of 2-amino-5-bromopyridine. J. Chem. Soc., Dalton Trans. 2003, 59 (5), 1113-1118.