m-Toluic acid, 99%
m-Toluic acid, 99%
m-Toluic acid, 99%
Thermo Scientific Chemicals
m-Toluic acid, 99%
CAS: 99-04-7 | C8H8O2 | 136.15 g/mol
Have Questions?
Catalog NumberQuantity
FSKA13785.0B1000 g
Catalog number FSKA13785.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or Materialm-Toluic acid
CAS99-04-7
Health Hazard 1H318
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P280-P305+P351+P338-P310
View more
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.
Solubility
Soluble in alcohol and ether.
Notes
Incompatible with strong oxidizing agents. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only
General References:
  1. Yong Ge; Frederic H Vaillancourt; Nathalie Y R Agar, Lindsay D Eltis. Reactivity of toluate dioxygenase with substituted benzoates and dioxygen. Journal of Bacteriology.2002, 184 (15), 4096-4103
  2. I Sarand, S Timonen, T Koivula, R Peltola, K Haahtela, R Sen, M Romantschuk. Tolerance and biodegradation of m-toluate by Scots pine, a mycorrhizal fungus and fluorescent pseudomonads individually and under associative conditions.Journal of Applied Microbiology.1999, 86 (5), 817-826