Sulfanilamide, 98%
Sulfanilamide, 98%
Sulfanilamide, 98%
Thermo Scientific Chemicals
Sulfanilamide, 98%
CAS: 63-74-1 | C6H8N2O2S | 172.202 g/mol
Have Questions?
Catalog NumberQuantity
FSKA13001.22100 g
Catalog number FSKA13001.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfanilamide
CAS63-74-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.
Solubility
Soluble in water, acetone, ethanol, glycerol, propylene glycol and hydrochloric acid. Insoluble in chloroform, ether, benzene and petroleum ether.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Intermediate for preparation of 2,6-disubstituted anilines, by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid. See, e.g.: Org. Synth. Coll., 3, 262 (1955).
  2. Yang, L.; Zhou, S.; Xiao, Y.; Tang, Y.; Xie, T. Sensitive simultaneous determination of three sulfanilamide artificial sweeters by capillary electrophoresis with on-line preconcentration and contactless conductivity detection. Food Chem. 2015, 188, 446-451.
  3. Bhattacharya, A.; Sharma, M.; Gulati, A.; Joshi, R., Chanda, S. K.; Ahuja, P. S. Histochemical evaluation of catechins in PEG stressed transgenic tea plants using catechin-specific-diazotized sulfanilamide reagent. Biotech. Histochem. 2015, 90 (1), 45-54.