L-Ornithine hydrochloride, 99%
L-Ornithine hydrochloride, 99%
L-Ornithine hydrochloride, 99%
L-Ornithine hydrochloride, 99%
Thermo Scientific Chemicals
L-Ornithine hydrochloride, 99%
CAS: 3184-13-2 | C5H13ClN2O2 | 168.621 g/mol
Have Questions?
Catalog NumberQuantity
FSKA12111.1425 g
Catalog number FSKA12111.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Ornithine hydrochloride
Melting Point∼238°C (decomposition)
CAS3184-13-2
Recommended StorageAmbient temperatures
RTECS NumberRM2985000
View more
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.
Solubility
Soluble in water (543 mg/ml at 20°C), and alcohol.
Notes
Store at room temperature. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. A. Chiba; T. Ueki; T. Ashida; Y. Sasada and M. Kakudo. The crystal structure of L-ornithine hydrochloride. Acta Cryst. 1967, 22 (6), 863-870.
  2. B. Dittrich; P. Munshi and M. A. Spackman. Redetermination, invariom-model and multipole refinement of L-ornithine hydrochloride. Acta Cryst. 2007, 63 (3), 505-509.