1-Octen-3-ol, 98%
1-Octen-3-ol, 98%
1-Octen-3-ol, 98%
Thermo Scientific Chemicals
1-Octen-3-ol, 98%
CAS: 3391-86-4 | C8H16O | 128.215 g/mol
Have Questions?
Catalog NumberQuantity
FSKA12079.1850 g
Catalog number FSKA12079.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Octen-3-ol
CAS3391-86-4
Health Hazard 1H227-H302-H315-H319-H332
Health Hazard 2GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
View more
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
RUO – Research Use Only
General References:
  1. Bo Sahlberg.; Maria Gunnbjörnsdottir.; Argo Soon.; Rain Jogi.; Thorarinn Gislason.; Gunilla Wieslander.; Christer Janson.; Dan Norback. Airborne molds and bacteria, microbial volatile organic compounds (MVOC), plasticizers and formaldehyde in dwellings in three North European cities in relation to sick building syndrome (SBS).Sci Total Environ.2013,444433-440.
  2. Viviana Polizzi.; An Adams.; Sarah De Saeger.; Carlos Van Peteghem.; Antonio Moretti.; Norbert De Kimpe.; Viviana Polizzi.; An Adams.; Sarah De Saeger.; Carlos Van Peteghem.; Antonio Moretti.; Norbert De Kimpe. Influence of various growth parameters on fungal growth and volatile metabolite production by indoor molds.Sci Total Environ.2012,414 277-286.