Sodium dihydrogen phosphate dihydrate, 98+%
Sodium dihydrogen phosphate dihydrate, 98+%
Sodium dihydrogen phosphate dihydrate, 98+%
Sodium dihydrogen phosphate dihydrate, 98+%
Thermo Scientific Chemicals
Sodium dihydrogen phosphate dihydrate, 98+%
CAS: 13472-35-0 | H6NaO6P | 156.006 g/mol
Have Questions?
Catalog NumberQuantity
FSKA11316.36500 g
Catalog number FSKA11316.36
Price (SGD)
102.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
102.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium dihydrogen phosphate dihydrate
CAS13472-35-0
Recommended StorageAmbient temperatures
RTECS NumberWA1900000
Quantity500 g
View more
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
Solubility
Soluble in water.(1 M at 20°C)
Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
RUO – Research Use Only
L Bohaty; S Haussühl. Elastic, electrostrictive, and electro-optic properties of orthorhombic sodium dihydrogen phosphate dihydrate. Zeitschrift für Kristallographie-Crystalline Materials.1984, 167, 303-306.