Indium(III) selenide, 99.99% (metals basis)
Indium(III) selenide, 99.99% (metals basis)
Indium(III) selenide, 99.99% (metals basis)
Indium(III) selenide, 99.99% (metals basis)
Thermo Scientific Chemicals
Indium(III) selenide, 99.99% (metals basis)
CAS: 12056-07-4 | In2Se3
Have Questions?
Catalog NumberQuantity
FSK088280.1425 g
Catalog number FSK088280.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIndium(III) selenide
Melting Point∽890°C
CAS12056-07-4
ColorSilver to Gray
Health Hazard 1H301+H331-H373
View more
Indium(III) selenide is used in photovoltaic devices. It is also used for optoelectronics and in solar cells.