Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis)
Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis)
Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis)
Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis)
Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis)
Thermo Scientific Chemicals
Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis)
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
FSK046986.RG30 x 150 cm
Catalog number FSK046986.RG
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
30 x 150 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper foil
CAS7440-50-8
Recommended StorageAmbient temperatures
RTECS NumberGL5325000
EINECS Number231-159-6