Iron(III) isopropoxide, 98%
Iron(III) isopropoxide, 98%
Iron(III) isopropoxide, 98%
Iron(III) isopropoxide, 98%
Thermo Scientific Chemicals
Iron(III) isopropoxide, 98%
CAS: 14995-22-3 | C9H21FeO3 | 233.109 g/mol
Have Questions?
Catalog NumberQuantity
FSK044873.065 g
Catalog number FSK044873.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6097-08-1
IUPAC Name2-(hex-5-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC14H13NO2
InChI KeyWALVSJIWVWDFCU-UHFFFAOYSA-N
SMILESO=C1N(CCCCC#C)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥96.0%
Iron(III) isopropoxide is applied in the field of nano-materials and thin films, engineering and environmental field. It is employed in catalysis and synthesis, energy conversion and storage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only