Aluminum oxide desiccant, regular
Aluminum oxide desiccant, regular
Aluminum oxide desiccant, regular
Aluminum oxide desiccant, regular
Thermo Scientific Chemicals
Aluminum oxide desiccant, regular
CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Catalog NumberQuantity
FSK044018.A32 kg
Catalog number FSK044018.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum oxide desiccant
Name NoteRegular
CAS1344-28-1
Recommended StorageAmbient temperatures
Physical FormSpheres
View more
Aluminum oxide is used in dessicators ceramics, coating material and electrical insulators. It is an ingredient in sunscreen, sometimes present in cosmetics such as blush, lipstick, and nail polish. Aluminum oxide is the catalyst in the Claus process in hydro desulfurization and some Ziegler-Natta polymerizations. It is widely used as an abrasive, in the automotive industries, cosmetic industries and fiber materials.It is a medium for chromatography in some sodium vapor lamps.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Dry gases and liquids
  • Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2
  • Liquids include: benzene, hexane, oil, pentane
RUO – Research Use Only