Polyethylene, low density, 1000 micron
Polyethylene, low density, 1000 micron
Polyethylene, low density, 1000 micron
Polyethylene, low density, 1000 micron
Thermo Scientific Chemicals
Polyethylene, low density, 1000 micron
CAS: 9002-88-4 | (C2H4)n | 28.05 g/mol
Have Questions?
Catalog NumberQuantity
FSK043949.A11 kg
Catalog number FSK043949.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene
Name NoteLow density, 1000 micron
CAS9002-88-4
Physical FormGranular
Recommended StorageAmbient temperatures
View more
Polyethylene is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for the manufacturing of various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipments. It is used in the manufacture of plastic bags, yard bags or outdoor bags, which is an environmentally sensible material.