Polyethylene glycol 8,000
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Thermo Scientific Chemicals
Polyethylene glycol 8,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Catalog NumberQuantity
FSK043443.22100 g
Catalog number FSK043443.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene glycol 8,000
ColorWhite
pH5 to 7
CAS25322-68-3
Physical FormPowder
View more
Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In gas chromatography, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies. In pharmaceutical industry, it is used as tablet and capsule lubricant.