Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Thermo Scientific Chemicals
Methylene Blue, 1% w/v aq. soln.
CAS: 122965-43-9 | C16H18ClN3S
Have Questions?
Catalog NumberQuantity
FSK042771.AE100 mL
Catalog number FSK042771.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS122965-43-9, 7732-18-5
Specifications
Chemical Name or MaterialMethylene Blue
CAS122965-43-9
CAS7732-18-5
Recommended StorageAmbient temperatures
TSCAYes
View more
Methylene Blue, 1% w/v aqueous solution is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It is also used in the sulfide analysis.