Tetrahydrofuran, anhydrous, 99.8+%, unstab., packaged under Argon in resealable ChemSeal™ bottles
For Synthesis and Liquid Chromatography | CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Catalog Number
Quantity
FSK041820.AK
250 mL
Catalog number FSK041820.AK
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrahydrofuran
Melting Point-108°C
Name NoteAnhydrous, UnStabilized
CAS109-99-9