Phthalic anhydride, ACS, 99.0-100.2%
Phthalic anhydride, ACS, 99.0-100.2%
Phthalic anhydride, ACS, 99.0-100.2%
Phthalic anhydride, ACS, 99.0-100.2%
Thermo Scientific Chemicals
Phthalic anhydride, ACS, 99.0-100.2%
CAS: 85-44-9 | C8H4O3 | 148.12 g/mol
Have Questions?
Catalog NumberQuantity
FSK041771.36500 g
Catalog number FSK041771.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhthalic anhydride
CAS85-44-9
Health Hazard 1H302-H315-H317-H318-H334-H335-H500
Health Hazard 2GHS H Statement
H334-H318-H302-H335-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
View more
Phthalic anhydride is used as an intermediate in chemical synthesis and plastics manufacturing.