Sodium oxalate, ACS, 99.5+%
Sodium oxalate, ACS, 99.5+%
Sodium oxalate, ACS, 99.5+%
Sodium oxalate, ACS, 99.5+%
Thermo Scientific Chemicals
Sodium oxalate, ACS, 99.5+%
CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol
Have Questions?
Catalog NumberQuantity
FSK041759.30250 g
Catalog number FSK041759.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium oxalate
Melting Point250°C to 270°C (decomposition)
CAS62-76-0
ColorWhite
Health Hazard 1H302+H312-H320
View more
Sodium oxalate is used as a reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.