Ammonium chloride, ACS, 99.5% min
Ammonium chloride, ACS, 99.5% min
Ammonium chloride, ACS, 99.5% min
Ammonium chloride, ACS, 99.5% min
Thermo Scientific Chemicals
Ammonium chloride, ACS, 99.5% min
CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Catalog NumberQuantity
FSK040193.A11 kg
Catalog number FSK040193.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium chloride
Melting Point340°C (Sublimation)
CAS12125-02-9
Health Hazard 1H302-H319-H335-H373
Health Hazard 3P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
View more
Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacturing of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent.