Indium(III) nitrate hydrate, 99.99% (metals basis)
Indium(III) nitrate hydrate, 99.99% (metals basis)
Indium(III) nitrate hydrate, 99.99% (metals basis)
Indium(III) nitrate hydrate, 99.99% (metals basis)
Thermo Scientific Chemicals
Indium(III) nitrate hydrate, 99.99% (metals basis)
CAS: 207398-97-8 | InN3O9 | 300.83 g/mol
Have Questions?
Catalog NumberQuantity
FSK040185.1850 g
Catalog number FSK040185.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIndium(III) nitrate hydrate
Melting Point100°C-H2O
CAS207398-97-8
ColorWhite
Health Hazard 1H272-H315-H319
View more
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.