Potassium bromide, FTIR Grade
Potassium bromide, FTIR Grade
Potassium bromide, FTIR Grade
Potassium bromide, FTIR Grade
Thermo Scientific Chemicals
Potassium bromide, FTIR Grade
CAS: 7758-02-3 | BrK | 119.00 g/mol
Have Questions?
Catalog NumberQuantity
FSK039794.1425 g
Catalog number FSK039794.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point1435°C
Chemical Name or MaterialPotassium Bromide
Melting Point730°C
Name NoteFTIR Grade
CAS7758-02-3
View more
Potassium bromide is used as a veterinary drug, as an antiepileptic medication for dogs. It used for removal of peripheral membrane proteins. Potassium bromide and sodium perborate play an important role for mild and regioselective oxidative bromination of anilines. Its Pellet is useful for fine structural characteristics by infared spectroscopy.