Caffeine, 99.7%
Caffeine, 99.7%
Caffeine, 99.7%
Thermo Scientific Chemicals
Caffeine, 99.7%
CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
Have Questions?
Catalog NumberQuantity
FSK039214.1425 g
Catalog number FSK039214.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCaffeine
CAS58-08-2
Health Hazard 1H302
Health Hazard 3P264b-P270-P301+P312-P330-P501c
Melting Point234°C to 236°C
View more
It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.