Alizarin Yellow R sodium salt
Alizarin Yellow R sodium salt
Alizarin Yellow R sodium salt
Alizarin Yellow R sodium salt
Thermo Scientific Chemicals
Alizarin Yellow R sodium salt
CAS: 1718-34-9 | C13H8N3NaO5 | 309.213 g/mol
Have Questions?
Catalog NumberQuantity
FSK038707.0910 g
Catalog number FSK038707.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAlizarin Yellow R sodium salt
CAS1718-34-9
ColorBrown to Yellow
Health Hazard 1H302
Health Hazard 3P264b-P270-P301+P312-P330-P501c