Phenolphthalein, ACS
Phenolphthalein, ACS
Phenolphthalein, ACS
Phenolphthalein, ACS
Thermo Scientific Chemicals
Phenolphthalein, ACS
Indicator | CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
Have Questions?
Catalog NumberQuantity
FSK038705.22100 g
Catalog number FSK038705.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenolphthalein
Melting Point258°C to 262°C
Name NoteFor analysis ACS
CAS77-09-8
Health Hazard 1H341-H351-H361f-H373
View more
Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extend of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.