Methyl Red, ACS
Methyl Red, ACS
Methyl Red, ACS
Thermo Scientific Chemicals
Methyl Red, ACS
CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol
Have Questions?
Catalog NumberQuantity
FSK036682.1425 g
Catalog number FSK036682.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl Red
Melting Point179°C to 182°C
Name NoteFor analysis ACS
CAS493-52-7
Health Hazard 1H373
View more
pH indicator in pH range 4.2 - 6.3Methyl red is used as a pH indicator. It is red in pH less than 4.4 and yellow in pH over 6.2. It is used to study its degradation using Fenton's reagent. In microbiology, it is used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose.