o-Tolidine dihydrochloride, ACS
o-Tolidine dihydrochloride, ACS
o-Tolidine dihydrochloride, ACS
o-Tolidine dihydrochloride, ACS
Thermo Scientific Chemicals
o-Tolidine dihydrochloride, ACS
CAS: 612-82-8 | C14H18Cl2N2 | 285.212 g/mol
Have Questions?
Catalog NumberQuantity
FSK036626.36500 g
Catalog number FSK036626.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Materialo-Tolidine dihydrochloride
CAS612-82-8
Health Hazard 1H302-H350
Health Hazard 2GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
Health Hazard 3P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
View more
o-Tolidine is employed as a colorimetric reagent for gold and free chlorine in water. Used in the synthesis of 9,9-bis(3-methyl-4-aminophenyl)fluorine. It is utilized for the detection of reducing sugars by thin layer chromatography. It is used in hair dyes, as a pharmaceutical intermediate and laboratory reagent.