Calcium carbonate, chelometric standard, ACS, 99.95-100.05%
Calcium carbonate, chelometric standard, ACS, 99.95-100.05%
Calcium carbonate, chelometric standard, ACS, 99.95-100.05%
Calcium carbonate, chelometric standard, ACS, 99.95-100.05%
Thermo Scientific Chemicals
Calcium carbonate, chelometric standard, ACS, 99.95-100.05%
Calcium carbonate, CCaO3, CAS Number-471-34-1, aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble, 500g, White powder, 99.95-100.05%, CHEBI:3311, 2.93, 207-439-9, 100.09, ACS, VTYYLEPIZMXCLO-UHFFFAOYSA-L, calcium;carbonate | CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Catalog NumberQuantity
FSK036399.22100 g
Catalog number FSK036399.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium carbonate
Melting Point800°C (decomposition)
Name NoteFor analysis ACS
CAS471-34-1
ColorWhite
View more
Calcium carbonate is a dietary supplement used when the quantity of calcium taken in the diet is not enough. Used as an antacid to relieve heartburn, acid indigestion and upset belly. Practiced in the expression industry. Also widely used as a filler in plastics and is used in metallurgy of iron.