Benzoic acid, ACS, 99.5% min
Benzoic acid, ACS, 99.5% min
Benzoic acid, ACS, 99.5% min
Benzoic acid, ACS, 99.5% min
Thermo Scientific Chemicals
Benzoic acid, ACS, 99.5% min
CAS: 65-85-0 | C7H6O2 | 122.123 g/mol
Have Questions?
Catalog NumberQuantity
FSK036230.1425 g
Catalog number FSK036230.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzoic acid
pH2.5 to 3.5
CAS65-85-0
Melting Point121°C to 124°C
Boiling Point249°C
View more
Benzoic acid is an important precursor for the preparation of many other organic substances viz. benzoyl chloride, benylbenzoate, phenol, sodium benzoate, benzyl alcohol as well as benzoate plasticizers such as glycol-, diethhyleneglycol- and triethyleneglycol esters. Benzoic acid and its salts are used as food preservatives. It is involved in rubber polymerization as an activator and a retardant. It is the main component of benzoin resin and is a constituent of Whitfield’s ointment, which is used for the treatment of fungal skin diseases such as tinea, ringworm and athlete’s foot. It is widely used in cosmetics, dyes, plastics and insect repellents.