Titanium foil, 0.032mm (0.0013in) thick, annealed, 99.7% (metals basis)
Titanium foil, 0.032mm (0.0013in) thick, annealed, 99.7% (metals basis)
Titanium foil, 0.032mm (0.0013in) thick, annealed, 99.7% (metals basis)
Titanium foil, 0.032mm (0.0013in) thick, annealed, 99.7% (metals basis)
Thermo Scientific Chemicals
Titanium foil, 0.032mm (0.0013in) thick, annealed, 99.7% (metals basis)
Form: Approx. 8.3g/23 x 25cm | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
FSK035881.WF23 x 25 cm
Catalog number FSK035881.WF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
23 x 25 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialTitanium
ColorSilver to Gray
Name NoteAnnealed
CAS7440-32-6
Melting Point1660°C