Cesium iodide, ultra dry, 99.998% (metals basis)
Cesium iodide, ultra dry, 99.998% (metals basis)
Cesium iodide, ultra dry, 99.998% (metals basis)
Cesium iodide, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals
Cesium iodide, ultra dry, 99.998% (metals basis)
CAS: 7789-17-5 | CsI | 259.81 g/mol
Have Questions?
Catalog NumberQuantity
FSK035729.1850 g
Catalog number FSK035729.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Boiling Point1280°C
Chemical Name or MaterialCesium iodide
Melting Point626°C
Name NoteUltra dry
CAS7789-17-5
View more
Cesium iodide is used as a beam splitter in Fourier transform infrared (FT-IR) spectrometers. It is widely used as a scintillator in silicon photodiodes. It is utilized in calorimeters and detectors and also employed as phosphor in fluoroscopy equipments. It finds application in making cell windows and prisms in infrared spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only