Sodium hydroxide, 10.0N Standardized Solution
Sodium hydroxide, 10.0N Standardized Solution
Sodium hydroxide, 10.0N Standardized Solution
Thermo Scientific Chemicals
Sodium hydroxide, 10.0N Standardized Solution
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Catalog NumberQuantity
FSK035635.K21 L
Catalog number FSK035635.K2
Price (SGD)
120.00
Each
Quantity:
1 L
Request bulk or custom format
Price (SGD)
120.00
Each
Specifications
Chemical Name or MaterialSodium hydroxide
ColorColorless
Name NoteStandardized Solution
CAS1310-73-2
Health Hazard 1H290-H314-H335
View more
Sodium hydroxide, 10.0N standardized solution is used for volumetric analysis in laboratories.