Potassium hydroxide, 10% w/v aq. soln.
Potassium hydroxide, 10% w/v aq. soln.
Potassium hydroxide, 10% w/v aq. soln.
Potassium hydroxide, 10% w/v aq. soln.
Thermo Scientific Chemicals
Potassium hydroxide, 10% w/v aq. soln.
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantity
FSK035615.K21 L
Catalog number FSK035615.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hydroxide
CAS1310-58-3
ColorColorless
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.