Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Thermo Scientific Chemicals
Silver nitrate, 0.1N Standardized Solution
CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Have Questions?
Catalog NumberQuantity
FSK035601.AP500 mL
Catalog number FSK035601.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver nitrate
CAS7761-88-8
ColorColorless
Health Hazard 1H315-H319
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.