ColorpHast™ pH Strips, Intermediate, three squares with different indicators on each strip, 7.5-14 pH range
ColorpHast™ pH Strips, Intermediate, three squares with different indicators on each strip, 7.5-14 pH range
Thermo Scientific Chemicals
ColorpHast™ pH Strips, Intermediate, three squares with different indicators on each strip, 7.5-14 pH range
Have Questions?
Catalog NumberpH RangeQuantity
FSK035237.LS0 to 146 Unit
Catalog number FSK035237.LS
View Price:Sign in to see your account pricing. Need an account? Register with us today.
pH Range:
0 to 14
Quantity:
6 Unit
Request bulk or custom format
Specifications
pH Range0 to 14
Quantity6 Unit
TypeIntermediate pH Strip
Unit SizeEA
ColorpHast™ pH Strips, Universal, four squares with different indicators on each strip, 0-14 pH range is used in analytical laboratory to measure the pH readings in seconds in drinking water, saliva and in chemical reactions. It is utilized for quick and accurate measurement and saves our time and trouble.