Magnesium sulfate, anhydrous, 99.5% min
Magnesium sulfate, anhydrous, 99.5% min
Magnesium sulfate, anhydrous, 99.5% min
Magnesium sulfate, anhydrous, 99.5% min
Thermo Scientific Chemicals
Magnesium sulfate, anhydrous, 99.5% min
CAS: 7487-88-9 | MgO4S | 120.36 g/mol
Have Questions?
Catalog NumberQuantity
FSK033337.36500 g
Catalog number FSK033337.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium sulfate, anhydrous
Melting Point1124°C (decomposition)
Name Noteanhydrous
CAS7487-88-9
Storage Note 1Hygroscopic
View more
Magnesium sulfate is suitable for sensitive molecular biology applications. It used as a component of bath salts and beauty product. It is utilized by the athletes to soothe sore muscles and effective in the removal of splinters. It is commonly used as a desiccant in organic synthesis due to its affinity for water. It acts as a brewing salt in beer production to adjust the ion content of the brewing water as well as enhance enzyme action. It plays a vital role as an antidote for barium chloride poisoning. . It is useful in controlling seizures associated with epilepsy, glomerulonephritis, or hypothyroidism.