Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
Thermo Scientific Chemicals
Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Catalog NumberQuantity
FSK033314.AP500 mL
Catalog number FSK033314.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialFormaldehyde, 37% in aq. soln.
Name NoteFor ACS analysis
CAS50-00-0
Health Hazard 1H226-H301+H311+H331-H314-H317-H335-H336-H341-H350-H370-H372
Health Hazard 2GHS H Statement
H301-H311-H331-H370-H351-H314-H226-H317
View more
Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and is used to make components for transmissions, electrical systems, engine blocks, door panels, axles and brake shoes. It serves as a disinfectant. It is also an ingredient in glues and is used as a preservative in medical laboratories, as an embalming fluid and as a sterilizer.