Cyclohexanone, ACS, 99+%
Cyclohexanone, ACS, 99+%
Cyclohexanone, ACS, 99+%
Cyclohexanone, ACS, 99+%
Thermo Scientific Chemicals
Cyclohexanone, ACS, 99+%
CAS: 108-94-1 | C6H10O | 98.145 g/mol
Have Questions?
Catalog NumberQuantity
FSK033309.K21 L
Catalog number FSK033309.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclohexanone
Melting Point°C to 26°C
Name NoteFor ACS analysis
CAS108-94-1
ColorColorless
View more
Cyclohexanone is used as an industrial solvent and paint remover. It acts as a precursor to nylon 6,6 and nylon 6 and cyclohexanone oxime, which gives caprolactam on rearrangement. Further, it is used as a chemical reaction medium, adhesives, sealants and agricultural products.