Sulfuric acid, ACS, 95.0-98.0%
Sulfuric acid, ACS, 95.0-98.0%
Sulfuric acid, ACS, 95.0-98.0%
Sulfuric acid, ACS, 95.0-98.0%
Thermo Scientific Chemicals
Sulfuric acid, ACS, 95.0-98.0%
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
FSK033273.A11 kg
Catalog number FSK033273.A1
Price (SGD)
152.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
152.00
EA
Add to cart
Specifications
Chemical Name or MaterialSulfuric Acid
CAS7664-93-9
Health Hazard 1H314-H330-H350
Density1.84
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
View more
Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.