Methanol, ACS, absolute, low acetone, 99.8+%
Methanol, ACS, absolute, low acetone, 99.8+%
Methanol, ACS, absolute, low acetone, 99.8+%
Methanol, ACS, absolute, low acetone, 99.8+%
Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantity
FSK031721.AP500 mL
Catalog number FSK031721.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMethanol
Melting Point°C to 98°C
CAS67-56-1
ColorColorless
Health Hazard 1H225-H301+H311+H331-H370-H372
View more
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin.